Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNC2 SIRNA Assays ›
Gene Symbol:
Tnnc2
Gene Name:
troponin C type 2 (fast)
Gene Aliases:
-
Chromosome Location:
Chr.3: 161243747 - 161244831 on Build Rnor_6.0
UniGene ID:
Rn.43529
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s150099
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037351.1
NP_001032428.1
3, 4
319
GenBank mRNA
DQ273679.1
-
3, 4
319
FQ215069.1
-
383
FQ216531.1
-
383
FQ216869.1
-
383
FQ216875.1
-
413
FQ216940.1
-
375
FQ216956.1
-
396
FQ217047.1
-
383
FQ217054.1
-
383
FQ217106.1
-
412
FQ217137.1
-
383
FQ217169.1
-
383
FQ217172.1
-
383
FQ217195.1
-
383
FQ217236.1
-
394
FQ217299.1
-
383
FQ217371.1
-
383
FQ217378.1
-
383
FQ217706.1
-
383
FQ217788.1
-
383
FQ217840.1
-
383
FQ217842.1
-
383
FQ217896.1
-
382
FQ217902.1
-
383
FQ217974.1
-
412
FQ217984.1
-
383
FQ218010.1
-
383
FQ218048.1
-
413
FQ218061.1
-
383
FQ223543.1
-
383
FQ223572.1
-
383
FQ223583.1
-
383
FQ223607.1
-
383
FQ223623.1
-
383
FQ223636.1
-
383
FQ223643.1
-
383
FQ223690.1
-
383
FQ223692.1
-
383
FQ223711.1
-
383
FQ223713.1
-
383
FQ223719.1
-
383
FQ223773.1
-
395
FQ223816.1
-
404
FQ223825.1
-
383
FQ223832.1
-
383
FQ223835.1
-
413
FQ223976.1
-
383
FQ223986.1
-
383
FQ224079.1
-
383
FQ224114.1
-
412
FQ224128.1
-
383
FQ224223.1
-
383
FQ224258.1
-
383
FQ224340.1
-
383
FQ224376.1
-
398
FQ224378.1
-
383
FQ224387.1
-
383
FQ224450.1
-
383
FQ224462.1
-
383
FQ224599.1
-
383
FQ224648.1
-
383
FQ224672.1
-
383
FQ224735.1
-
384
FQ224749.1
-
383
FQ224755.1
-
412
FQ224795.1
-
383
FQ224822.1
-
383
FQ224827.1
-
380
J00793.1
-
1, 2
103
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
actin binding
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-dvnl5:80/10.239.162.83:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0