Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199491.1 | NP_955785.1 | 3 | 934 | ||
XM_006233585.2 | XP_006233647.1 | 2 | 820 | |||
XM_006233586.2 | XP_006233648.1 | 2 | 514 | |||
XM_006233587.2 | XP_006233649.1 | 2 | 543 | |||
XM_008761582.1 | XP_008759804.1 | 2 | 618 | |||
XM_008761583.1 | XP_008759805.1 | 2 | 769 | |||
GenBank mRNA | AJ276068.2 | - | 3 | 1034 |