Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106579.1 | NP_001100049.1 | 5, 6 | 1146 | ||
XM_008762698.1 | XP_008760920.1 | 4, 5 | 914 |