Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrguk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106589.1 | NP_001100059.1 | 14 | 1702 | ||
XM_006236253.2 | XP_006236315.1 | 14 | 1716 | |||
XM_006236255.2 | XP_006236317.1 | 14 | 1716 | |||
XM_006236256.2 | XP_006236318.1 | 14 | 1716 | |||
XM_008762766.1 | XP_008760988.1 | 14 | 1713 |