Search Thermo Fisher Scientific
Gene Symbol: Rad52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106617.2 | NP_001100087.2 | 7 | 573 | ||
XM_008763232.1 | XP_008761454.1 | 8 | 1310 | |||
XM_008763233.1 | XP_008761455.1 | 8 | 1315 | |||
XM_008763234.1 | XP_008761456.1 | 8 | 1311 | |||
XM_008763235.1 | XP_008761457.1 | 8 | 1310 | |||
XM_008763236.1 | XP_008761458.1 | 8 | 1309 | |||
XM_008763237.1 | XP_008761459.1 | 8 | 1310 | |||
XM_008763238.1 | XP_008761460.1 | 8 | 1310 |