Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009660.1 | NP_001009660.1 | 2 | 262 | ||
XM_006237336.2 | XP_006237398.1 | 2 | 751 | |||
XM_006237337.2 | XP_006237399.1 | 2 | 298 | |||
GenBank mRNA | BC088189.1 | - | 2 | 262 | 7371446 |
Gene Symbol: LOC100911548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750655.3 | XP_003750703.1 | 2 | 374 | ||
XM_006244625.2 | XP_006244687.1 | 2 | 747 | |||
XM_006244626.2 | XP_006244688.1 | 2 | 296 |