Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2j1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106642.1 | NP_001100112.1 | 3 | 192 | ||
XM_008763612.1 | XP_008761834.1 | 3 | 290 | |||
XM_008763613.1 | XP_008761835.1 | 3 | 290 |