Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106656.1 | NP_001100126.1 | 4 | 590 | ||
XM_006238035.2 | XP_006238097.1 | 4 | 706 | |||
XM_006238036.2 | XP_006238098.1 | 4 | 1625 | |||
XM_008763697.1 | XP_008761919.1 | 4 | 706 |