Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007131.4 | NP_009062.2 | 5 | 3334 | ||
XM_005262469.1 | XP_005262526.1 | 5 | 1863 | |||
XM_005262471.3 | XP_005262528.1 | 3 | 2309 | |||
XM_011531391.1 | XP_011529693.1 | 6 | 2050 | |||
XM_011531396.2 | XP_011529698.1 | 4 | 2827 | |||
GenBank mRNA | AK092347.1 | - | 2 | 223 | ||
AK290215.1 | - | 4 | 1038 | |||
AL831984.2 | - | 5 | 3279 | |||
AL832801.1 | - | 4 | 821 | |||
BC109100.2 | - | 4 | 853 | 40034784 | ||
S67970.1 | - | 1 | 59 |