Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185063.2 | NP_001171992.1 | 4 | 1149 | ||
NM_007131.4 | NP_009062.2 | 7 | 3754 | |||
XM_005262469.1 | XP_005262526.1 | 7 | 2283 | |||
XM_005262471.3 | XP_005262528.1 | 5 | 2729 | |||
XM_011531391.1 | XP_011529693.1 | 8 | 2470 | |||
XM_011531395.2 | XP_011529697.1 | 4 | 2883 | |||
XM_011531396.2 | XP_011529698.1 | 6 | 3247 | |||
XM_011531397.2 | XP_011529699.1 | 5 | 3155 | |||
XM_017029818.1 | XP_016885307.1 | 6 | 3684 | |||
XM_017029819.1 | XP_016885308.1 | 4 | 6449 | |||
XM_017029820.1 | XP_016885309.1 | 5 | 1662 | |||
GenBank mRNA | AK091687.1 | - | 4 | 1143 | ||
AK092347.1 | - | 4 | 643 | |||
AK290215.1 | - | 6 | 1458 | |||
AL831984.2 | - | 7 | 3699 | |||
AL832801.1 | - | 6 | 1241 | |||
BC109100.2 | - | 6 | 1273 | 40034784 | ||
S67970.1 | - | 2 | 817 |