Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302109.1 | NP_001289038.1 | 7 | 1530 | ||
NM_001324041.1 | NP_001310970.1 | 7 | 1487 | |||
NM_001324085.1 | NP_001311014.1 | 5 | 1223 | |||
NM_153028.3 | NP_694573.1 | 6 | 1006 | |||
NR_126069.1 | 5 | 1165 | ||||
NR_136681.1 | 6 | 1334 | ||||
NR_136683.1 | 5 | 810 | ||||
NR_136686.1 | 6 | 1360 | ||||
XM_011522646.2 | XP_011520948.1 | 6 | 1419 | |||
XM_011522647.2 | XP_011520949.1 | 6 | 1104 | |||
XM_017023642.1 | XP_016879131.1 | 6 | 902 | |||
GenBank mRNA | AK056091.1 | - | 6 | 929 | ||
AK093979.1 | - | 5 | 1118 | |||
BC119652.1 | - | 5 | 731 | 40115880 | ||
BC119653.1 | - | 5 | 731 | 40115881 | ||
CR749782.1 | - | 6 | 829 |