Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199253.2 | NP_954862.2 | 8 | 1440 | ||
GenBank mRNA | AY847775.1 | - | 8 | 1440 | ||
BC133063.1 | - | 8 | 1371 | 8371686 |