Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191585.1 | NP_001178514.1 | 7 | 1688 | ||
XM_006257447.2 | XP_006257509.1 | 8 | 1892 |