Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047868.1 | NP_001041333.1 | 5 | 232 | ||
XM_006238796.2 | XP_006238858.1 | 4 | 247 | |||
XM_006238797.2 | XP_006238859.1 | 4 | 256 | |||
GenBank mRNA | AY325152.1 | - | 5 | 232 | ||
BC158560.1 | - | 4 | 197 | 9102303 |