Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129996.1 | NP_001123468.1 | 4 | 1099 | ||
NM_013360.2 | NP_037492.2 | 4 | 1067 | |||
XM_005259208.3 | XP_005259265.1 | 5 | 1163 | |||
XM_005259209.3 | XP_005259266.1 | 3 | 1020 | |||
GenBank mRNA | AF187988.1 | - | 4 | 1046 | ||
AK095196.1 | - | 6 | 1259 | |||
BC030261.1 | - | 4 | 1041 | 5210539 |
Gene Symbol: ZNF223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF187989.1 | - | 5 | 1215 |