Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029917.1 | NP_001025088.1 | 6 | 669 | ||
GenBank mRNA | BC098061.1 | - | 6 | 669 | 7458219 | |
FQ215386.1 | - | 4 | 421 |