Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106751.2 | NP_001100221.1 | 1 | 515 | ||
XM_008764761.1 | XP_008762983.1 | 2 | 1023 | |||
GenBank mRNA | EF405799.1 | - | 1 | 515 |