Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137633.1 | NP_001131105.1 | 13 | 2290 | ||
XM_006240577.2 | XP_006240639.1 | 12 | 2298 | |||
GenBank mRNA | FQ212109.1 | - | 12 | 2048 |