Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001380.1 | NP_001001380.1 | 1 | 64 |
Gene Symbol: Olr1091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000593.1 | NP_001000593.1 | 1 | 64 |
Gene Symbol: Olr1095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000416.1 | NP_001000416.1 | 1 | 64 |
Set Membership: |
G-protein coupled receptor |