Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106775.1 | NP_001100245.1 | 2 | 342 | ||
XM_006241197.1 | XP_006241259.1 | 2 | 247 | |||
XM_006241198.2 | XP_006241260.1 | 2 | 242 | |||
GenBank mRNA | BC091253.1 | - | 1 | 104 | ||
BC161937.1 | - | 2 | 214 | 9022830 | ||
FQ220712.1 | - | 2 | 225 |