Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106781.2 | NP_001100251.2 | 2 | 162 | ||
GenBank mRNA | BF555697.1 | - | 2 | 137 | ||
BG671355.1 | - | 1 | 13 | |||
FQ212338.1 | - | 2 | 147 | |||
FQ217295.1 | - | 2 | 148 | |||
FQ224477.1 | - | 2 | 148 |
Gene Symbol: LOC100910710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239763.2 | XP_006239825.1 | 2 | 159 |