Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127527.2 | NP_001120999.1 | 7 | 641 | ||
GenBank mRNA | BC166753.1 | - | 7 | 641 | 5623659 |