Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004239.1 | NP_001004239.1 | 7 | 723 | ||
XM_006241448.1 | XP_006241510.1 | 8 | 811 | |||
XM_008765392.1 | XP_008763614.1 | 8 | 813 | |||
GenBank mRNA | BC079042.1 | - | 7 | 723 | 7114929 |