Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013104.1 | NP_001013122.1 | 2 | 1141 | ||
XM_008765549.1 | XP_008763771.1 | 2 | 3205 | |||
XM_008765550.1 | XP_008763772.1 | 2 | 1266 | |||
XM_008765551.1 | XP_008763773.1 | 4 | 1258 | |||
XM_008765552.1 | XP_008763774.1 | 4 | 1239 | |||
XM_008765553.1 | XP_008763775.1 | 2 | 1158 | |||
XM_008765554.1 | XP_008763776.1 | 2 | 1269 | |||
XM_008765555.1 | XP_008763777.1 | 2 | 1269 | |||
XM_008765556.1 | XP_008763778.1 | 2 | 1269 | |||
XM_008765557.1 | XP_008763779.1 | 2 | 145 | |||
XM_008765558.1 | XP_008763780.1 | 2 | 152 | |||
XM_008765559.1 | XP_008763781.1 | 2 | 132 | |||
XM_008765560.1 | XP_008763782.1 | 2 | 145 | |||
XM_008765561.1 | XP_008763783.1 | 2 | 145 | |||
XM_008765562.1 | XP_008763784.1 | 2 | 130 | |||
GenBank mRNA | BC079391.1 | - | 2 | 1141 | 7111852 | |
FQ220813.1 | - | 2 | 102 | |||
FQ225933.1 | - | 2 | 137 | |||
FQ227443.1 | - | 2 | 113 | |||
FQ233760.1 | - | 2 | 93 |