Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYM2 SIRNA Assays ›
Gene Symbol:
ZMYM2
Gene Name:
zinc finger MYM-type containing 2
Gene Aliases:
FIM, MYM, RAMP, SCLL, ZNF198
Chromosome Location:
Chr.13: 19957412 - 20091845 on Build GRCh38
UniGene ID:
Hs.507433
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15266
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190964.2
NP_001177893.1
14
2575
NM_001190965.2
NP_001177894.1
13
2590
NM_003453.4
NP_003444.1
14
2683
NM_197968.3
NP_932072.1
13
2636
XM_005266520.3
XP_005266577.1
13
2595
XM_011535216.2
XP_011533518.1
14
2265
XM_011535222.1
XP_011533524.1
13
2526
XM_011535223.2
XP_011533525.1
7
4980
XM_011535224.2
XP_011533526.1
7
881
XM_017020728.1
XP_016876217.1
13
2526
XM_017020729.1
XP_016876218.1
13
2501
XM_017020731.1
XP_016876220.1
14
2318
XM_017020732.1
XP_016876221.1
14
2340
XM_017020733.1
XP_016876222.1
15
2367
XM_017020734.1
XP_016876223.1
15
2353
XM_017020735.1
XP_016876224.1
14
2317
XM_017020736.1
XP_016876225.1
14
2489
XM_017020737.1
XP_016876226.1
14
2264
XM_017020738.1
XP_016876227.1
13
2579
GenBank mRNA
AF012126.1
-
7
978
AF035374.1
-
11
1881
AF060181.1
-
11
1933
AJ224901.1
-
14
2569
AK226118.1
-
13
2570
AK302917.1
-
8
950
BC036372.1
-
13
2636
4823008
BX647944.1
-
13
2590
BX648905.1
-
6
3020
Y13472.1
-
14
2581
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
biological_process
peptidyl-tyrosine phosphorylation
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein tyrosine kinase activity
zinc ion binding
ubiquitin conjugating enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-q5jv4:80/10.239.161.22:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0