Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008324.1 | NP_001008325.1 | 10 | 1283 | ||
GenBank mRNA | BC085933.1 | - | 10 | 1283 | 7112408 |