Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750589.3 | XP_003750637.1 | 6, 7 | 2038 |
Gene Symbol: Atrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106859.1 | NP_001100329.1 | 6, 7 | 1043 | ||
XM_006243764.2 | XP_006243826.1 | 6, 7 | 1045 | |||
XM_006243766.2 | XP_006243828.1 | 6, 7 | 1044 | |||
XM_006243767.2 | XP_006243829.1 | 6, 7 | 1044 |