Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdspl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106865.1 | NP_001100335.1 | 3 | 261 | ||
XM_008766675.1 | XP_008764897.1 | 1 | 56 |