Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198130.1 | NP_937763.1 | 5 | 842 | ||
XM_006244087.2 | XP_006244149.1 | 6 | 1189 | |||
GenBank mRNA | AY191270.1 | - | 5 | 842 | ||
BC097266.1 | - | 5 | 782 | 7459961 |