Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106883.1 | NP_001100353.1 | 2 | 694 | ||
GenBank mRNA | BC168177.1 | - | 2 | 681 | 9035251 | |
FQ214885.1 | - | 2 | 696 | |||
FQ214919.1 | - | 2 | 586 | |||
FQ215260.1 | - | 2 | 696 | |||
FQ216053.1 | - | 2 | 696 | |||
FQ216724.1 | - | 2 | 693 | |||
FQ216808.1 | - | 2 | 582 | |||
FQ217810.1 | - | 2 | 696 | |||
FQ224967.1 | - | 2 | 729 |