Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106887.1 | NP_001100357.1 | 4 | 682 | ||
XM_006244428.2 | XP_006244490.1 | 3 | 637 |