Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191812.1 | NP_001178741.1 | 9 | 2635 | ||
XM_006245376.2 | XP_006245438.1 | 9 | 3168 |