Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286457.1 | NP_001273386.1 | 15 | 1690 | ||
NM_001286458.1 | NP_001273387.1 | 16 | 1865 | |||
NM_001321858.1 | NP_001308787.1 | 15 | 1616 | |||
NM_003470.2 | NP_003461.2 | 15 | 1821 | |||
XM_017023652.1 | XP_016879141.1 | 15 | 2249 | |||
XM_017023653.1 | XP_016879142.1 | 15 | 1685 | |||
GenBank mRNA | AK302481.1 | - | 16 | 1791 | ||
AK302771.1 | - | 15 | 1622 | |||
AK302912.1 | - | 16 | 1809 | |||
AK316441.1 | - | 15 | 1691 | |||
AY376241.1 | - | 15 | 1652 | |||
Z72499.1 | - | 15 | 1821 |