Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005551.2 | NP_001005551.1 | 6 | 787 | ||
XM_006256859.2 | XP_006256921.1 | 7 | 884 | |||
GenBank mRNA | BC081872.1 | - | 6 | 789 | 7106842 |