Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106970.1 | NP_001100440.1 | 1, 2 | 221 | ||
GenBank mRNA | BC168927.1 | - | 1, 2 | 142 | 7108747 |