Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001657.3 | NP_001648.1 | 4 | 832 | ||
GenBank mRNA | BC009799.1 | - | 4 | 798 | 4277616 | |
BC146953.1 | - | 4 | 639 | 9056881 | ||
BC146967.1 | - | 4 | 639 | 9056895 | ||
BT019866.1 | - | 4 | 584 | |||
M30704.1 | - | 4 | 793 |