Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pwwp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127296.1 | NP_001120768.1 | 4 | 1815 | ||
XM_006246139.2 | XP_006246201.1 | 3 | 2606 | |||
GenBank mRNA | BC088289.1 | - | 1 | 150 | ||
BC166858.1 | - | 4 | 1815 | 9100439 | ||
FQ234972.1 | - | 2 | 326 |