Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106995.1 | NP_001100465.1 | 2 | 309 | ||
GenBank mRNA | FQ211728.1 | - | 2 | 313 |