Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107079.1 | NP_001100549.1 | 4 | 1116 | ||
XM_006248699.2 | XP_006248761.1 | 4 | 1458 | |||
XM_006248700.2 | XP_006248762.1 | 4 | 1458 | |||
XM_006248701.2 | XP_006248763.1 | 5 | 1345 |