Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Filip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248213.2 | XP_006248275.2 | 2 | 4906 | ||
XM_008768652.1 | XP_008766874.1 | 4 | 3249 | |||
XM_008768653.1 | XP_008766875.1 | 5 | 1297 | |||
XM_008768654.1 | XP_008766876.1 | 4 | 3249 | |||
XM_008768655.1 | XP_008766877.1 | 4 | 3249 | |||
XM_008768656.1 | XP_008766878.1 | 4 | 3249 | |||
XM_008768657.1 | XP_008766879.1 | 2 | 445 |