Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Brwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751022.3 | XP_003751070.1 | 11, 12 | 1203 | ||
XM_006248185.2 | XP_006248247.1 | 11, 12 | 1203 | |||
XM_006248186.2 | XP_006248248.1 | 11, 12 | 1206 |