Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107131.1 | NP_001100601.1 | 3 | 505 | ||
XM_006249140.2 | XP_006249202.1 | 3 | 528 | |||
XM_006249141.2 | XP_006249203.1 | 3 | 527 | |||
XM_006249142.2 | XP_006249204.1 | 3 | 529 |