Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013438.2 | NP_001013456.1 | 15 | 1806 | ||
NM_001013439.2 | NP_001013457.1 | 16 | 2150 | |||
NM_005087.3 | NP_005078.2 | 15 | 1806 | |||
XM_005247813.3 | XP_005247870.2 | 14 | 1668 | |||
XM_005247814.3 | XP_005247871.2 | 14 | 1668 | |||
XM_005247815.3 | XP_005247872.2 | 15 | 1581 | |||
XM_005247816.3 | XP_005247873.2 | 14 | 1668 | |||
XM_006713775.3 | XP_006713838.1 | 15 | 2011 | |||
XM_017007298.1 | XP_016862787.1 | 15 | 2011 | |||
XM_017007299.1 | XP_016862788.1 | 16 | 1924 | |||
XM_017007300.1 | XP_016862789.1 | 15 | 2011 | |||
XM_017007301.1 | XP_016862790.1 | 16 | 1924 | |||
GenBank mRNA | AK225334.1 | - | 15 | 1582 | ||
AK292633.1 | - | 15 | 1582 | |||
AK294603.1 | - | 14 | 1528 | |||
AK297254.1 | - | 13 | 1434 | |||
AK297331.1 | - | 13 | 1574 | |||
AK302197.1 | - | 15 | 1585 | |||
AY341428.1 | - | 15 | 1857 | |||
BC028983.1 | - | 15 | 1576 | 5122574 | ||
KR259877.1 | - | 15 | 1566 | |||
KR259878.1 | - | 14 | 1653 | |||
KR259879.1 | - | 14 | 1653 | |||
KR259880.1 | - | 16 | 1878 | |||
U25165.1 | - | 15 | 1578 |