Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107165.1 | NP_001100635.1 | 2, 3 | 684 | ||
XM_008772381.1 | XP_008770603.1 | 3, 4 | 808 | |||
XM_008772382.1 | XP_008770604.1 | 3, 4 | 823 | |||
XM_008772383.1 | XP_008770605.1 | 3, 4 | 805 | |||
XM_008772384.1 | XP_008770606.1 | 3, 4 | 782 | |||
XM_008772385.1 | XP_008770607.1 | 4, 5 | 744 | |||
XM_008772386.1 | XP_008770608.1 | 4, 5 | 619 | |||
XM_008772387.1 | XP_008770609.1 | 3, 4 | 637 | |||
XM_008772388.1 | XP_008770610.1 | 3, 4 | 754 | |||
XM_008772389.1 | XP_008770611.1 | 3, 4 | 731 | |||
XM_008772390.1 | XP_008770612.1 | 3, 4 | 750 | |||
XM_008772391.1 | XP_008770613.1 | 3, 4 | 735 | |||
XM_008772392.1 | XP_008770614.1 | 3, 4 | 449 | |||
XM_008772393.1 | XP_008770615.1 | 3, 4 | 758 | |||
XM_008772394.1 | XP_008770616.1 | 3, 4 | 538 | |||
XM_008772395.1 | XP_008770617.1 | 3, 4 | 531 | |||
XM_008772396.1 | XP_008770618.1 | 3, 4 | 611 | |||
XM_008772397.1 | XP_008770619.1 | 3, 4 | 564 | |||
XM_008772398.1 | XP_008770620.1 | 3, 4 | 512 |