Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT88 SIRNA Assays ›
Gene Symbol:
IFT88
Gene Name:
intraflagellar transport 88
Gene Aliases:
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location:
Chr.13: 20566446 - 20691437 on Build GRCh38
UniGene ID:
Hs.187376
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15623
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT88
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318493.1
NP_001305422.1
5
960
NM_006531.4
NP_006522.2
4
632
NM_175605.4
NP_783195.2
6
861
NR_134653.1
5
960
XM_005266553.2
XP_005266610.1
6
1222
XM_006719870.3
XP_006719933.1
4
327
XM_011535241.2
XP_011533543.1
5
435
XM_011535242.1
XP_011533544.1
7
1362
XM_011535243.1
XP_011533545.1
6
1241
XM_017020757.1
XP_016876246.1
5
1340
XM_017020759.1
XP_016876248.1
5
893
XM_017020760.1
XP_016876249.1
6
794
XM_017020761.1
XP_016876250.1
5
430
XM_017020762.1
XP_016876251.1
5
893
XM_017020763.1
XP_016876252.1
5
893
XM_017020764.1
XP_016876253.1
4
339
XM_017020765.1
XP_016876254.1
4
339
XM_017020768.1
XP_016876257.1
5
893
XM_017020770.1
XP_016876259.1
6
794
XM_017020771.1
XP_016876260.1
6
1222
XM_017020772.1
XP_016876261.1
4
339
XM_017020774.1
XP_016876263.1
4
285
XM_017020775.1
XP_016876264.1
5
893
XM_017020776.1
XP_016876265.1
5
893
GenBank mRNA
AK126668.1
-
5
1331
AK303853.1
-
4
286
AK308281.1
-
4
287
AK310223.1
-
6
517
BC030776.1
-
6
542
4824965
U20362.1
-
4
376
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cilium morphogenesis
regulation of cilium assembly
regulation of autophagosome assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0