Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107171.1 | NP_001100641.1 | 2 | 222 | ||
XM_006249686.2 | XP_006249748.1 | 2 | 230 | |||
XM_008769457.1 | XP_008767679.1 | 2 | 230 |