Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107177.1 | NP_001100647.1 | 23 | 3928 | ||
XM_008769555.1 | XP_008767777.1 | 23 | 3940 |