Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107192.1 | NP_001100662.1 | 7 | 1282 | ||
XM_006250112.2 | XP_006250174.1 | 7 | 1292 | |||
XM_008769649.1 | XP_008767871.1 | 7 | 1385 | |||
GenBank mRNA | AY724478.1 | - | 7 | 1253 |