Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100683.1 | NP_001094153.1 | 2 | 1069 | ||
GenBank mRNA | CK471950.1 | - | 2 | 331 |