Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC14A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF349446.1 | - | 1207 | |||
AK074236.1 | - | 4 | 491 | |||
AK122698.1 | - | 8 | 1063 | |||
AK292422.1 | - | 5 | 712 | |||
BC110445.1 | - | 3 | 415 | |||
BC110446.1 | - | 3 | 415 | 40008423 |